
Calpain Inhibitor II

Synonym(s):
N-Acetyl-L-leucyl-L-leucyl-L-methioninal
Empirical Formula (Hill Notation):
C19H35N3O4S
CAS Number:
Molecular Weight:
401.56
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1