Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Calcium fluoride
Synonym(s):
Fluorite, Fluorspar
Empirical Formula (Hill Notation):
CaF
2
CAS Number:
7789-75-5
Molecular Weight:
78.07
Products
Product Category
salts (1)
Brand
Sigma-Aldrich (3)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
449717
Calcium fluoride,
anhydrous, powder, 99.99% trace metals basis
Expand
View Pricing
378801
Calcium fluoride,
random crystals, optical grade, 99.99% trace metals basis
Expand
View Pricing
1.02840
Calcium fluoride,
precipitated pure, Technipur
®
Expand
View Pricing