Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Calcium D-saccharate tetrahydrate
Synonym(s):
D-Glucaric acid calcium salt, D-Saccharic acid calcium salt, Calcium D-glucarate
Empirical Formula (Hill Notation):
C
6
H
8
CaO
8
· 4H
2
O
CAS Number:
5793-89-5
Molecular Weight:
320.26
EC Number:
227-334-1
Products
Product Category
carbohydrates (1)
disaccharides (1)
Biological Source
Color
Formula Weight
Physical Form
Purity
Optical Isomer
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
21236
Calcium D-saccharate tetrahydrate,
98.5-102% (KT)
Expand
View Pricing