
Butyl 2-methylbutyrate

Linear Formula:
CH3CH2CH(CH3)COOCH2CH2CH2CH3
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1