Bupivacaine Related Compound B
Synonym(s):
Desbutylbupivacaine, (RS)-N-(2,6-Dimethylphenyl)piperidine-2-carboxamide
Empirical Formula (Hill Notation):
C14H20N2O
CAS Number:
Molecular Weight:
232.32
Products
Brand
Formula Weight
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing