Skip to Content
Merck
Bumetanide British Pharmacopoeia (BP) Reference Standard

Bumetanide

Bumetanide British Pharmacopoeia (BP) Reference Standard
Synonym(s):
3-(Aminosulfonyl)-5-(butylamino)-4-phenoxybenzoic acid
Empirical Formula (Hill Notation):
C17H20N2O5S
CAS Number:
Molecular Weight:
364.42
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade