Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Buffer solution
Products
Brand
Supelco (42)
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
certified reference material (38)
Filter Products
Showing 1-20 of 45
Page 1 of 3
Page 1 of 3
Compare
Product Number
Description
Pricing
1.09439
Buffer solution,
(di-sodium hydrogen phosphate/potassium dihydrogen phosphate), traceable to SRM from NIST and PTB pH 7.00 (20°C) Certipur
®
Expand
View Pricing
1.09435
Buffer solution,
(citric acid/sodium hydroxide/hydrogen chloride), traceable to SRM from NIST and PTB pH 4.00 (20°C) Certipur
®
Expand
View Pricing
1.09438
Buffer solution,
(boric acid/potassium chloride/sodium hydroxide), traceable to SRM from NIST and PTB pH 10.00 (20°C) Certipur
®
Expand
View Pricing
1.09475
Buffer solution,
(citric acid/sodium hydroxide/hydrogen chloride) colour: red traceable to SRM from NIST and PTB pH 4.00 (20 °C) Certipur
®
Expand
View Pricing
1.09477
Buffer solution,
(di-sodium hydrogen phosphate/potassium dihydrogen phosphate), colour: green traceable to SRM from NIST and PTB pH 7.00 (20°C) Certipur
®
Expand
View Pricing
1.09461
Buffer solution,
(boric acid/potassium chloride/sodium hydroxide), traceable to SRM from NIST and PTB pH 9.00 (20°C) Certipur
®
Expand
View Pricing
1.09433
Buffer solution,
(citric acid/sodium hydroxide/hydrogen chloride), traceable to SRM from NIST and PTB pH 2.00 (20°C) Certipur
®
Expand
View Pricing
1.09407
Buffer solution,
(potassium dihydrogen phosphate/disodium hydrogen phosphate)traceable to SRM from NIST and PTB pH 7.00 (25°C) Certipur
®
Expand
View Pricing
1.09409
Buffer solution,
(boric acid/potassium chloride/sodium hydroxide)traceable to SRM from NIST and PTB pH 10.00 (25°C) Certipur
®
Expand
View Pricing
1.09406
Buffer solution,
(potassium hydrogen phthalate), traceable to SRM from NIST and PTB pH 4.01 (25°C) Certipur
®
Expand
View Pricing
1.99002
Buffer solution,
(potassium dihydrogen phosphate/di-sodium hydrogen phosphate), traceable to SRM from NIST and PTB pH 7.00 (25°C) Certipur
®
Expand
View Pricing
1.09400
Buffer solution,
(boric acid/potassium chloride/sodium hydroxide) colour coded: yellow, tracable to NIST and PTB pH 10.00 (20°C) Certipur
®
Expand
View Pricing
1.09476
Buffer solution,
(boric acid/potassium chloride/sodium hydroxide), coloured: blue traceable to SRM from NIST and PTB pH 9.00 (20°C) Certipur
®
Expand
View Pricing
1.99004
Buffer solution,
(boric acid/potassium chloride/sodium hydroxide), traceable to SRM from NIST and PTB pH 10.00 (25°C) Certipur
®
Expand
View Pricing
1.09445
Buffer solution,
(citric acid/sodium hydroxide/hydrogen chloride) traceable to SRM from NIST and PTB pH 4.00 (25°C) Certipur
®
Expand
View Pricing
1.99001
Buffer solution,
(potassium hydrogen phthalate), traceable to SRM from NIST and PTB pH 4.01 (25°C) Certipur
®
Expand
View Pricing
1.99022
Buffer solution,
(di-sodium hydrogen phosphate/sodium hydroxide) traceable to SRM from NIST und PTB pH 12.00 (25°C) Certipur
®
Expand
View Pricing
1.99054
Buffer solution,
(citric acid/sodium hydroxide/hydrogen chloride) colour: red, traceable to SRM from NIST and PTB pH 4.00 (25 °C), Certipur
®
Expand
View Pricing
1.99012
Buffer solution,
(citric acid, sodium hydroxide, hydrogen chloride) tracable to SRM from NIST and PTB pH 2.00 (25°C) Certipur
®
Expand
View Pricing
1.99057
Buffer solution,
(di-sodium hydrogen phosphate/potassium dihydrogen phosphate), colour:yellowtraceable to SRM from NIST and PTB pH 7.00 (25°C) Certipur
®
Expand
View Pricing
Page 1 of 3
Page 1 of 3