Skip to Content
Products
Cart
196
AR
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
196
Back
Buffer concentrate
Products
Product Category
buffers (10)
Brand
Supelco (13)
pH Value
Product Line
Agency
Format
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (11)
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
1.09892
Buffer concentrate,
(phosphate/sodium hydroxide), traceable to SRM of NIST and PTB for 500 ml buffer solution, pH 12.00 ± 0.05 (20°C) Titrisol
®
Expand
View Pricing
1.09887
Buffer concentrate,
(phosphate), traceable to SRM of NIST and PTB for 500 ml buffer solution, pH 7.00 ± 0.02 (20°C) Titrisol
®
Expand
View Pricing
1.09889
Buffer concentrate,
(boric acid/potassium chloride/sodium hydroxide), traceable to SRM of NIST and PTB for 500 ml buffer solution, pH 9.00 ± 0.02 (20°C) Titrisol
®
Expand
View Pricing
1.09883
Buffer concentrate,
(citrate/hydrochloric acid), traceable to SRM of NIST and PTB for 500 ml buffer solution, pH 3.00 ± 0.02 (20°C) Titrisol
®
Expand
View Pricing
1.09882
Buffer concentrate,
(citrate/hydrochloric acid), traceable to SRM of NIST and PTB for 500 ml buffer solution, pH 2.00 ± 0.02 (20°C) Titrisol
®
Expand
View Pricing
1.09884
Buffer concentrate,
(citrate/hydrochloric acid), traceable to SRM of NIST and PTB for 500 ml buffer solution, pH 4.00 ± 0.02 (20°C) Titrisol
®
Expand
View Pricing
1.09890
Buffer concentrate,
(boric acid/potassium chloride/sodium hydroxide), traceable to SRM of NIST and PTB for 500 ml buffer solution, pH 10.00 ± 0.05 (20°C) Titrisol
®
Expand
View Pricing
1.09893
Buffer concentrate,
(potassium chloride/sodium hydroxide), traceable to SRM of NIST and PTB for 500 ml buffer solution, pH 13.00 ± 0.05 (20°C) Titrisol
®
Expand
View Pricing
1.09881
Buffer concentrate,
(glycine/hydrochloric acid), traceable to SRM of NIST and PTB for 500 ml buffer solution, pH 1.00 ± 0.02 (20°C) Titrisol
®
Expand
View Pricing
1.09888
Buffer concentrate,
(borate/hydrochloric acid), traceable to SRM of NIST and PTB for 500 ml buffer solution, pH 8.00 ± 0.02 (20°C) Titrisol
®
Expand
View Pricing
1.09880
Buffer concentrate,
(boric acid/potassium chloride/sodium hydroxide), traceable to SRM of NIST and PTB for 500 ml buffer solution, pH 11.00 ± 0.05 (20°C) Titrisol
®
Expand
View Pricing
1.09885
Buffer concentrate,
(citrate/sodium hydroxide), traceable to SRM of NIST and PTB for 500 ml buffer solution, pH 5.00 ± 0.02 (20°C) Titrisol
®
Expand
View Pricing
1.09886
Buffer concentrate,
(citrate/sodium hydroxide), traceable to SRM of NIST and PTB for 500 ml buffer solution, pH 6.00 ± 0.02 (20°C) Titrisol
®
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
Español - ES
English - EN
Learn More