Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
spare burette tip
Products
Product Category
burettes (4)
Manufacturer
Volume (ml)
Color
Package Size
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
BR11511
BRAND
®
spare burette tip,
for burette lengths 25 mL, clear borosilicate glass 3.3
Expand
View Pricing
BR11510
BRAND
®
spare burette tip,
for burette lengths 10 mL, clear borosilicate glass 3.3
Expand
View Pricing
BR11512
BRAND
®
spare burette tip,
for burette lengths 50 mL, clear borosilicate glass 3.3
Expand
View Pricing
BR11515
BRAND
®
spare burette tip,
for compact/automatic burette 25+50 mL, amber glass
Expand
View Pricing