Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
bottle for dispensers
Products
Product Category
bottles (10)
Manufacturer
Capacity (mL)
Diameter (mm)
Height (mm)
Cap
Bottle material
Color
Vial Bottle Material
Vial Bottle Color
Package Size
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
BR704018
BRAND
®
bottle for dispensers,
capacity 1000 mL, without coating
Expand
View Pricing
BR704008
BRAND
®
bottle for dispensers,
capacity 1000 mL, with Ethyl-Acrylate coat
Expand
View Pricing
BR704020
BRAND
®
bottle for dispensers,
capacity 2500 mL, without coating
Expand
View Pricing
BR704010
BRAND
®
bottle for dispensers,
capacity 2500 mL, with Ethyl-Acrylate coat
Expand
View Pricing
BR704016
BRAND
®
bottle for dispensers,
capacity 500 mL, without coating
Expand
View Pricing
BR704004
BRAND
®
bottle for dispensers,
capacity 250 mL, with Ethyl-Acrylate coat
Expand
View Pricing
BR704012
BRAND
®
bottle for dispensers,
capacity 100 mL, without coating
Expand
View Pricing
BR704014
BRAND
®
bottle for dispensers,
capacity 250 mL, without coating
Expand
View Pricing
BR704006
BRAND
®
bottle for dispensers,
capacity 500 mL, with Ethyl-Acrylate coat
Expand
View Pricing
BR704002
BRAND
®
bottle for dispensers,
capacity 100 mL, with Ethyl-Acrylate coat
Expand
View Pricing