Skip to Content
Merck
Boc-Val-OH ≥99.0% (T)

Boc-Val-OH

Boc-Val-OH ≥99.0% (T)
Synonym(s):
(S)-2-(Boc-amino)-3-methylbutyric acid, N-(tert-Butoxycarbonyl)-L-valine, Boc-L-valine, Boc-Val-OH
Linear Formula:
(CH3)2CHCH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
217.26

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Reagent Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing