Skip to Content
Merck
Bis(tetrabutylammonium) dichromate 99%

Bis(tetrabutylammonium) dichromate

Bis(tetrabutylammonium) dichromate 99%
Synonym(s):
TBADC, Tetrabutylammonium dichromate
Linear Formula:
[(CH3CH2CH2CH2)4N]2Cr2O7
CAS Number:
Molecular Weight:
700.92
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing