Skip to Content
Merck
Bisdesmethoxycurcumin certified reference material, pharmaceutical secondary standard

Bisdesmethoxycurcumin

Bisdesmethoxycurcumin certified reference material, pharmaceutical secondary standard
Synonym(s):
1,7-Bis(4-hydroxyphenyl)-1,6-heptadiene-3,5-dione
Empirical Formula (Hill Notation):
C19H16O4
CAS Number:
Molecular Weight:
308.33

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade