![Bisabolol oxide A primary reference standard](/deepweb/assets/sigmaaldrich/product/structures/404/373/095a90f9-e4d3-47fb-bf4f-79c682b69fb4/640/095a90f9-e4d3-47fb-bf4f-79c682b69fb4.png)
Bisabolol oxide A
![Bisabolol oxide A primary reference standard](/deepweb/assets/sigmaaldrich/product/structures/404/373/095a90f9-e4d3-47fb-bf4f-79c682b69fb4/640/095a90f9-e4d3-47fb-bf4f-79c682b69fb4.png)
Synonym(s):
(3S,6S)-Tetrahydro-2,2,6-trimethyl-6-[(1S)-4-methyl-3-cyclohexen-1-yl]-2H-pyran-3-ol
Empirical Formula (Hill Notation):
C15H26O2
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing