Skip to Content
Merck
Biotin-16-ddUTP ≥98% (HPLC), solution, 1 mM, suitable for hybridization

Biotin-16-ddUTP

Biotin-16-ddUTP ≥98% (HPLC), solution, 1 mM, suitable for hybridization
Synonym(s):
Bio-16-ddUTP tetralithium salt, 5-(N-[N-Biotinyl-ε-aminocaproyl-γ-aminobutyryl]-3-aminoallyl)-2′,3′-dideoxyuridine 5′-triphosphate
Empirical Formula (Hill Notation):
C32H52N7O17P3S
CAS Number:
Molecular Weight:
931.78

Products

Formula Weight
Manufacturer
Physical Form
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing