Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bilirubin
Empirical Formula (Hill Notation):
C
33
H
36
N
4
O
6
CAS Number:
635-65-4
Molecular Weight:
584.66
Products
Product Category
bioactive small molecules (1)
Brand
Sigma-Aldrich (2)
Formula Weight
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
Special Grade
Purum (1)
metrological institute (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
B4126
Bilirubin,
≥98% (EmM/453 = 60), powder
Expand
View Pricing
14370
Bilirubin,
purum, ≥95.0% (UV)
Expand
View Pricing
NIST916B
Bilirubin,
NIST
®
SRM
®
Expand
View Pricing