Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Big CHAP - CAS 86303-22-2 - Calbiochem
Synonym(s):
BigCHAP, N,N-Bis[3-(D-gluconamido)propyl]cholamide
Empirical Formula (Hill Notation):
C
42
H
75
N
3
O
16
CAS Number:
86303-22-2
Molecular Weight:
878.06
Products
Brand
Millipore (1)
Formula Weight
Quality Segment
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
200965
Big CHAP - CAS 86303-22-2 - Calbiochem,
Non-ionic detergent analog of CHAPS and CHAPSO with significantly reduced electrostatic interactions that do not interfere in anion exchange chromatography on DEAE-cellulose. Aggregation number: 10.
Expand
View Pricing