Skip to Content
Merck
Benzo[ghi]perylene BCR®, certified reference material

Benzo[ghi]perylene

Benzo[ghi]perylene BCR®, certified reference material
Synonym(s):
1,12-Benzoperylene
Empirical Formula (Hill Notation):
C22H12
CAS Number:
Molecular Weight:
276.33

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade
Technique