Skip to Content
Merck
Benzo[a]pyrene solution 100 μg/mL in cyclohexane, analytical standard

Benzo[a]pyrene solution

Benzo[a]pyrene solution 100 μg/mL in cyclohexane, analytical standard
Empirical Formula (Hill Notation):
C20H12
CAS Number:
Molecular Weight:
252.31

Products

Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique