Skip to Content
Merck
Benserazide hydrochloride European Pharmacopoeia (EP) Reference Standard

Benserazide hydrochloride

Benserazide hydrochloride European Pharmacopoeia (EP) Reference Standard
Synonym(s):
DL-Serine 2-(2,3,4-trihydroxybenzyl)hydrazide hydrochloride
Empirical Formula (Hill Notation):
C10H15N3O5 · HCl
CAS Number:
Molecular Weight:
293.70

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Special Grade