Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Benchmark Orbi-Shaker
™
CO2 with remote controller
Synonym(s):
orbital shaker, microplate shaker, carbon dioxide incubator, CO2 incubation, harsh enviroment incubation
Products
Manufacturer
Length (cm)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
BMSBT4500EUK
Benchmark Orbi-Shaker
™
CO2 with remote controller,
AC/DC input 230 V, UK plug
Expand
View Pricing
BMSBT4500E
Benchmark Orbi-Shaker
™
CO2 with remote controller,
AC/DC input 230 V, EuroPlug
Expand
View Pricing
BMSBT4500
Benchmark Orbi-Shaker
™
CO2 with remote controller,
AC/DC input 115 V
Expand
View Pricing