Skip to Content
Merck
Bay 11-7082 ≥98% (HPLC), powder

Bay 11-7082

Bay 11-7082 ≥98% (HPLC), powder
Synonym(s):
(E)-3-(4-Methylphenylsulfonyl)-2-propenenitrile
Empirical Formula (Hill Notation):
C10H9NO2S
CAS Number:
Molecular Weight:
207.25

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing