Skip to Content
Merck
Bathophenanthrolinedisulfonic acid disodium salt hydrate 98%

Bathophenanthrolinedisulfonic acid disodium salt hydrate

Bathophenanthrolinedisulfonic acid disodium salt hydrate 98%
Synonym(s):
Bathophenanthrolinedisulfonic acid disodium salt trihydrate, 4,7-Diphenyl-1,10-phenanthroline-disulfonic acid disodium salt trihydrate, Disodium bathophenanthrolinedisulfonate trihydrate
Empirical Formula (Hill Notation):
C24H14N2Na2O6S2 · 3H2O
CAS Number:
Molecular Weight:
590.53
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing