Bathocuproinedisulfonic acid disodium salt
Synonym(s):
2,9-Dimethyl-4,7-diphenyl-1,10-phenantrolinedisulfonic acid disodium salt, Disodium bathocuproinedisulfonate, Bathocuproinedisulfonic acid disodium salt
Empirical Formula (Hill Notation):
C26H18N2Na2O6S2
CAS Number:
Molecular Weight:
564.54
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Application
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing