Skip to Content
Merck
Barium hydroxide octahydrate ACS reagent, ≥98%

Barium hydroxide octahydrate

Barium hydroxide octahydrate ACS reagent, ≥98%
Synonym(s):
Barium hydroxide octahydrate
Linear Formula:
Ba(OH)2 · 8 H2O
CAS Number:
Molecular Weight:
315.46

Products

Product Category
Brand
Sustainability
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Special Grade
Showing 1-5 of 5