
Azobisisobutyronitrile

Synonym(s):
2,2′-Azobis(2-methylpropionitrile) solution, α,α,′-Azoisobutyronitrile solution, AIBN solution
Empirical Formula (Hill Notation):
C8H12N4
CAS Number:
Molecular Weight:
164.21
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1