![Azathioprine Related Compound A](/deepweb/assets/sigmaaldrich/product/images/325/218/960c6826-37a7-4f5a-89f0-a01325ed81dd/640/960c6826-37a7-4f5a-89f0-a01325ed81dd.jpg)
Azathioprine Related Compound A
![Azathioprine Related Compound A](/deepweb/assets/sigmaaldrich/product/images/325/218/960c6826-37a7-4f5a-89f0-a01325ed81dd/640/960c6826-37a7-4f5a-89f0-a01325ed81dd.jpg)
Synonym(s):
1-Methyl-4-nitro-1H-imidazol-5-amine
Empirical Formula (Hill Notation):
C4H6N4O2
CAS Number:
Molecular Weight:
142.12
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing