
Azacitidine Related Compound B

Synonym(s):
1-O-Acetyl-2,3,5-tri-O-benzoyl-β-D-ribofuranose, β-D-Ribofuranose 1-acetate 2,3,5-tribenzoate
Empirical Formula (Hill Notation):
C28H24O9
CAS Number:
Molecular Weight:
504.48
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing