Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aurintricarboxylic acid
Synonym(s):
ATA
Empirical Formula (Hill Notation):
C
22
H
14
O
9
CAS Number:
4431-00-9
Molecular Weight:
422.34
EC Number:
224-628-1
Products
Product Category
inhibitors (1)
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Special Grade
Practical (1)
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
A1895
Aurintricarboxylic acid,
≥85% purity (titration), practical grade, powder
Expand
View Pricing