![Astragaloside IV European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/205/580/b3273ed4-4692-4939-ab73-56245c69ccea/640/b3273ed4-4692-4939-ab73-56245c69ccea.png)
Astragaloside IV
![Astragaloside IV European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/205/580/b3273ed4-4692-4939-ab73-56245c69ccea/640/b3273ed4-4692-4939-ab73-56245c69ccea.png)
Empirical Formula (Hill Notation):
C41H68O14
CAS Number:
Molecular Weight:
784.97
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade