![Aripiprazole Related Compound H Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/224/636/d82a88b5-9182-48e7-aa1d-f236d29e9292/640/d82a88b5-9182-48e7-aa1d-f236d29e9292.jpg)
Aripiprazole Related Compound H
![Aripiprazole Related Compound H Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/224/636/d82a88b5-9182-48e7-aa1d-f236d29e9292/640/d82a88b5-9182-48e7-aa1d-f236d29e9292.jpg)
Synonym(s):
7-(4-{4-[4-(2,3-Dichlorophenyl)piperazin-1-yl]butoxy}butoxy)-3,4-dihydroquinolin-2(1H)-one
Empirical Formula (Hill Notation):
C27H35Cl2N3O3
Molecular Weight:
520.49
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing