Skip to Content
Merck
Arabinose United States Pharmacopeia (USP) Reference Standard

Arabinose

Arabinose United States Pharmacopeia (USP) Reference Standard
Synonym(s):
L-(+)-Arabinose
Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13

Products

Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing