Skip to Content
AR
EN
Back
Apiin
Synonym(s):
Apigenin-7-(2-O-apiosylglucoside)
Empirical Formula (Hill Notation):
C
26
H
28
O
14
CAS Number:
26544-34-3
Molecular Weight:
564.49
EC Number:
247-780-0
Products
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
PHL89161
Apiin,
phyproof
®
Reference Substance
Expand
View Pricing