![α-Phenyl-2-pyridineacetamide certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/104/473/9340b780-9ddf-470e-aa3c-9f5e8177ba74/640/9340b780-9ddf-470e-aa3c-9f5e8177ba74.png)
α-Phenyl-2-pyridineacetamide
![α-Phenyl-2-pyridineacetamide certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/104/473/9340b780-9ddf-470e-aa3c-9f5e8177ba74/640/9340b780-9ddf-470e-aa3c-9f5e8177ba74.png)
Synonym(s):
Phenyl-(2-pyridyl)acetamide, α-Phenyl-2-pyridineacetamide
Empirical Formula (Hill Notation):
C13H12N2O
CAS Number:
Molecular Weight:
212.25
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing