Anti-β-Tubulin III Antibody
Synonym(s):
Anti-Tuj 1
Products
Product Category
Brand
Biological Source
Clonality
Color
Feature
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Color
Package Size
Post Translational Modification
Non-reactive Species
Species Reactivity
Technique
Showing 1-9 of 9
Compare
Product Number
Description
Pricing