Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Anti-Serotonin Transporter Antibody
Products
Product Category
antibodies (4)
primary antibodies (4)
Brand
Sigma-Aldrich (4)
Biological Source
Clonality
Gene Alias
Isotype
Antibody Form
Quality Segment
Post Translational Modification
Non-reactive Species
Species Reactivity
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
MAB5618
Anti-Serotonin Transporter Antibody,
ascites fluid, Chemicon
®
Expand
View Pricing
AB9726
Anti-Serotonin Transporter Antibody,
serum, Chemicon
®
Expand
View Pricing
AB10514P
Anti-Serotonin Transporter Antibody,
1 mg/mL, from rabbit
Expand
View Pricing
AB1594P
Anti-Serotonin Transporter Antibody,
Chemicon
®
, from rabbit
Expand
View Pricing