Skip to Content
Merck
Anti-MEK1 antibody produced in rabbit

Anti-MEK1 antibody produced in rabbit

Anti-MEK1 antibody produced in rabbit

Products

Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing