Anti-CD8B antibody produced in rabbit
Synonym(s):
CD8B1, LYT3, Leu2, Ly3, MGC119115
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Antibody Form
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing