Skip to Content
Merck
Anti-β-Tubulin III antibody produced in rabbit affinity isolated antibody, buffered aqueous solution

Anti-β-Tubulin III antibody produced in rabbit

Anti-β-Tubulin III antibody produced in rabbit affinity isolated antibody, buffered aqueous solution
Synonym(s):
Anti-Tuj 1

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Antibody Form
Conjugate
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1