Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Anthralin (Dithranol)
Synonym(s):
Dithranol, 1,8,9-Anthracenetriol, 1,8-Dihydroxy-9(10H)-anthracenone, 1,8-Dihydroxyanthrone, Anthralin
Empirical Formula (Hill Notation):
C
14
H
10
O
3
CAS Number:
1143-38-0
Molecular Weight:
226.23
EC Number:
214-538-0
Products
Brand
Supelco (1)
Formula Weight
Melting Point (°C)
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
certified reference material (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
PHR1677
Anthralin (Dithranol),
Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing