Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ampicillin
Synonym(s):
Ampicillin trihydrate, D-(−)-α-Aminobenzylpenicillin
Empirical Formula (Hill Notation):
C
16
H
19
N
3
O
4
S · 3H
2
O
CAS Number:
7177-48-2
Molecular Weight:
403.45
EC Number:
200-709-7
Products
Product Category
antibiotics (1)
Brand
Sigma-Aldrich (1)
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
Activity Spectrum
Agency
Quality Segment
Application
Mode of Action
Optical Isomer
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
A1593
Ampicillin,
meets USP testing specifications
Expand
View Pricing