Skip to Content
Merck
Ammonium tetrachlorocuprate(II) dihydrate puriss., ≥99.0% (AT)

Ammonium tetrachlorocuprate(II) dihydrate

Ammonium tetrachlorocuprate(II) dihydrate puriss., ≥99.0% (AT)
Synonym(s):
Ammonium chlorocuprate(II), Ammonium copper chloride
Linear Formula:
(NH4)2CuCl4 · 2H2O
CAS Number:
Molecular Weight:
277.47

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Reagent Type
Core
Special Grade
Showing 1-2 of 2