Skip to Content
Products
Cart
0
AR
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Ammonium fluoride
Synonym(s):
Ammonium fluoride
Linear Formula:
NH
4
F
CAS Number:
12125-01-8
Molecular Weight:
37.04
EC Number:
235-185-9
Products
Product Category
salts (2)
Brand
Sigma-Aldrich (2)
Supelco (1)
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Special Grade
ACS reagent (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
216011
Ammonium fluoride,
ACS reagent, ≥98.0%
Expand
View Pricing
1.01164
Ammonium fluoride,
for analysis EMSURE
®
ACS
Expand
View Pricing
338869
Ammonium fluoride,
≥99.99% trace metals basis
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
Español - ES
English - EN
Learn More