![Ametoctradin hydrochloride Metabolite M650F01 certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/190/463/a0cc2b01-25aa-49d0-8cb9-6bf6ce833333/640/a0cc2b01-25aa-49d0-8cb9-6bf6ce833333.png)
Ametoctradin hydrochloride Metabolite M650F01
![Ametoctradin hydrochloride Metabolite M650F01 certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/190/463/a0cc2b01-25aa-49d0-8cb9-6bf6ce833333/640/a0cc2b01-25aa-49d0-8cb9-6bf6ce833333.png)
Synonym(s):
4-(7-Amino-5-ethyl-[1,2,4]triazolo[1,5-a]pyrimidin-6-yl)butanoic acid hydrochloride, Propyl [3-(dimethylamino)propyl]carbamate, Propyl hydrogen [3-(dimethylamino)propyl]carbonimidate
Empirical Formula (Hill Notation):
C11H15N5O2 · HCl
CAS Number:
Molecular Weight:
285.73
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing