AmberChrom™ 1X4 Ion Exchange Resin
Linear Formula:
(C10H12 · C10H10 · C8H8 · C3H9N)X
CAS Number:
Products
Product Category
Brand
pH Value
Physical Form
Separation Technique
Quality Segment
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing