Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum-silicon-magnesium-manganese alloy, Al97.5Si1.0Mg0.8Mn0.7
Products
Manufacturer
Physical Form
Diameter (mm)
Length (mm)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
GF00128034
Aluminum-silicon-magnesium-manganese alloy, Al97.5Si1.0Mg0.8Mn0.7,
100mm rod, 9.5mm diameter, extruded
Expand
View Pricing
GF00480653
Aluminum-silicon-magnesium-manganese alloy, Al97.5Si1.0Mg0.8Mn0.7,
1000mm rod, 9.5mm diameter, extruded
Expand
View Pricing
GF16003842
Aluminum-silicon-magnesium-manganese alloy, Al97.5Si1.0Mg0.8Mn0.7,
500mm rod, 9.5mm diameter, extruded
Expand
View Pricing