Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum-magnesium alloy, Al95Mg5
Products
Manufacturer
Physical Form
Diameter (mm)
ID (mm)
Length (m)
Length (mm)
OD (mm)
Filter Products
Showing 1-20 of 21
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF36282056
Aluminum-magnesium alloy, Al95Mg5,
1000mm tube, 0.61mm outside diameter, 0.07mm wall thickness, 0.47mm inside diameter, as drawn
Expand
View Pricing
GF65460595
Aluminum-magnesium alloy, Al95Mg5,
500mm tube, 1.73mm outside diameter, 0.075mm wall thickness, 1.58mm inside diameter, as drawn
Expand
View Pricing
GF82573836
Aluminum-magnesium alloy, Al95Mg5,
1000mm tube, 1.73mm outside diameter, 0.075mm wall thickness, 1.58mm inside diameter, as drawn
Expand
View Pricing
GF17846556
Aluminum-magnesium alloy, Al95Mg5,
100mm tube, 0.75mm outside diameter, 0.125mm wall thickness, 0.5mm inside diameter, hard
Expand
View Pricing
GF28290258
Aluminum-magnesium alloy, Al95Mg5,
500mm tube, 0.61mm outside diameter, 0.07mm wall thickness, 0.47mm inside diameter, as drawn
Expand
View Pricing
GF16315411
Aluminum-magnesium alloy, Al95Mg5,
100mm tube, 0.61mm outside diameter, 0.07mm wall thickness, 0.47mm inside diameter, as drawn
Expand
View Pricing
GF09847389
Aluminum-magnesium alloy, Al95Mg5,
100mm tube, 1.73mm outside diameter, 0.075mm wall thickness, 1.58mm inside diameter, as drawn
Expand
View Pricing
GF30923681
Aluminum-magnesium alloy, Al95Mg5,
200mm tube, 1.73mm outside diameter, 0.075mm wall thickness, 1.58mm inside diameter, as drawn
Expand
View Pricing
GF59278775
Aluminum-magnesium alloy, Al95Mg5,
1000mm tube, 0.75mm outside diameter, 0.125mm wall thickness, 0.5mm inside diameter, hard
Expand
View Pricing
GF30819797
Aluminum-magnesium alloy, Al95Mg5,
200mm tube, 0.75mm outside diameter, 0.125mm wall thickness, 0.5mm inside diameter, hard
Expand
View Pricing
GF66542777
Aluminum-magnesium alloy, Al95Mg5,
1000mm tube, 0.74mm outside diameter, 0.063mm wall thickness, 0.614mm inside diameter, as drawn
Expand
View Pricing
GF07159451
Aluminum-magnesium alloy, Al95Mg5,
200mm tube, 0.74mm outside diameter, 0.063mm wall thickness, 0.614mm inside diameter, as drawn
Expand
View Pricing
GF52474237
Aluminum-magnesium alloy, Al95Mg5,
500mm tube, 0.75mm outside diameter, 0.125mm wall thickness, 0.5mm inside diameter, hard
Expand
View Pricing
GF35257271
Aluminum-magnesium alloy, Al95Mg5,
100mm tube, 0.74mm outside diameter, 0.063mm wall thickness, 0.614mm inside diameter, as drawn
Expand
View Pricing
GF39426329
Aluminum-magnesium alloy, Al95Mg5,
200mm tube, 0.61mm outside diameter, 0.07mm wall thickness, 0.47mm inside diameter, as drawn
Expand
View Pricing
GF94932344
Aluminum-magnesium alloy, Al95Mg5,
500mm tube, 0.74mm outside diameter, 0.063mm wall thickness, 0.614mm inside diameter, as drawn
Expand
View Pricing
GF12739563
Aluminum-magnesium alloy, Al95Mg5,
20m wire, 0.4mm diameter, annealed
Expand
View Pricing
GF22515971
Aluminum-magnesium alloy, Al95Mg5,
2m wire, 0.4mm diameter, annealed
Expand
View Pricing
GF40504407
Aluminum-magnesium alloy, Al95Mg5,
1m wire, 0.4mm diameter, annealed
Expand
View Pricing
GF66297246
Aluminum-magnesium alloy, Al95Mg5,
10m wire, 0.4mm diameter, annealed
Expand
View Pricing
Page 1 of 2
Page 1 of 2