Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Aluminum-cobalt alloy, Al99.9Co0.1
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
GF09427798
Aluminum-cobalt alloy, Al99.9Co0.1,
0.5m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF83624238
Aluminum-cobalt alloy, Al99.9Co0.1,
10m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF06717540
Aluminum-cobalt alloy, Al99.9Co0.1,
2m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF54923294
Aluminum-cobalt alloy, Al99.9Co0.1,
1m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF82920326
Aluminum-cobalt alloy, Al99.9Co0.1,
0.1m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF65861961
Aluminum-cobalt alloy, Al99.9Co0.1,
5m wire, 0.25mm diameter, as drawn
Expand
View Pricing
GF81341209
Aluminum-cobalt alloy, Al99.9Co0.1,
0.5m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF20210768
Aluminum-cobalt alloy, Al99.9Co0.1,
0.1m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF80852272
Aluminum-cobalt alloy, Al99.9Co0.1,
5m wire, 1.0mm diameter, as drawn
Expand
View Pricing
GF76381139
Aluminum-cobalt alloy, Al99.9Co0.1,
1m wire, 1.0mm diameter, as drawn
Expand
View Pricing