![Allopurinol Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/320/926/0c0203b0-fd85-4595-bd4a-1eeb1ee3d25f/640/0c0203b0-fd85-4595-bd4a-1eeb1ee3d25f.png)
Allopurinol Related Compound C
![Allopurinol Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/320/926/0c0203b0-fd85-4595-bd4a-1eeb1ee3d25f/640/0c0203b0-fd85-4595-bd4a-1eeb1ee3d25f.png)
Synonym(s):
3-(4H-1,2,4-Triazol-4-yl)-1H-pyrazole-4-carboxamide, 5-(4H-1,2,4-Triazol-4-yl)-1H-pyrazole-4-carboxamide
Empirical Formula (Hill Notation):
C6H6N6O
CAS Number:
Molecular Weight:
178.15
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2