![Agaric acid analytical standard](/deepweb/assets/sigmaaldrich/product/structures/314/864/425adaa0-03c5-441e-ba13-48bfb596315b/640/425adaa0-03c5-441e-ba13-48bfb596315b.png)
Agaric acid
![Agaric acid analytical standard](/deepweb/assets/sigmaaldrich/product/structures/314/864/425adaa0-03c5-441e-ba13-48bfb596315b/640/425adaa0-03c5-441e-ba13-48bfb596315b.png)
Synonym(s):
2-Hydroxy-1,2,3-nonadecanetricarboxylic acid, Agaricic acid, Agaricin
Empirical Formula (Hill Notation):
C22H40O7
CAS Number:
Molecular Weight:
416.55
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing